missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic acid, 98+%

Product Code. 11473194
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473194 500 g 500g
11448880 5 g 5g
11453194 25 g 25g
11463194 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11473194 Supplier Thermo Scientific Alfa Aesar Supplier No. A10556.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-51-4 | C10H9NO2 | 175.19 g/mol

It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.

Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

Melting Point ∼167°C (decomposition)
Flash Point 171°C (339°F)
Quantity 500 g
Beilstein 143358
Sensitivity Light sensitive
Merck Index 14,4964
Solubility Information Soluble in ethanol (50mg/ml),methanol,DMSO,and chloroform (sparingly). Insoluble in water.
Formula Weight 175.19
Percent Purity ≥98%
Chemical Name or Material Indole-3-acetic acid
compliance-icons
Product Identifier
  • Indole-3-acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Indole-3-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.