missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxylic Acid, TRC

Product Code. 30396040
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396040 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396040 Supplier Toronto Research Chemicals Supplier No. TRCI6270001G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1477-50-5
Molecular Formula C9 H7 N O2
Quantity 1 g
Synonym 1H-Indole-2-carboxylic acid, Indole-2-carboxylic acid (7CI,8CI), 2-Carboxyindole, 2-Indolylformic acid, NSC 16598
InChI Formula InChI=1S/C9H7NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h1-5,10H,(H,11,12)
SMILES OC(=O)c1cc2ccccc2[nH]1
IUPAC Name 1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 161.16
Formula Weight 161.0477
Chemical Name or Material Indole-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.