missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxaldehyde, 97%

Product Code. 15400878
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400878 1 g 1g
15420878 250 mg 250mg
15410878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400878 Supplier Thermo Scientific Alfa Aesar Supplier No. H54638.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19005-93-7 | C9H7NO | 145.16 g/mol

Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

Solubility
Soluble in Methanol.

Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19005-93-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD03001425
InChI Key SBNOTUDDIXOFSN-UHFFFAOYSA-N
Synonym indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, indole-2-carbaldehyde, 2-formylindole, 2-formyl-1h-indole, 1h-indolecarboxaldehyde, unii-r5n9cd7dlm, r5n9cd7dlm, methanol, 2h-indol-2-ylidene-, 1e-9ci, indolaldehyd
PubChem CID 96389
IUPAC Name 1H-indole-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 138°C to 142°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in Methanol.
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material Indole-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.