missing translation for 'onlineSavingsMsg'
Learn More

Indole-2,4,5,6,7-d5-3-acetic-Alpha,Alpha-d2 Acid, TRC

Product Code. 30400891
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400891 10 mg -
30404736 1 mg 1mg
30397955 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400891 Supplier Toronto Research Chemicals Supplier No. TRCI57734110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173020-21-7
Molecular Formula C10 2H7 H2 N O2
Molecular Weight (g/mol) 182.23
Synonym 2,2-Dideuterio-2-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)acetic acid, 1H-Indole-3-acetic acid D7, 1H-Indole-2,4,5,6,7-d5-3-acetic-α,α-d2 acid, 2-(3-Indolyl-2,4,5,6,7-d5)acetic-α,α-d2 acid, 3-Indolylacetic acid-d7
IUPAC Name 2,2-dideuterio-2-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)acetic acid
SMILES [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1C([2H])([2H])C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)/i1D,2D,3D,4D,5D2,6D
Formula Weight 182.1073
Chemical Name or Material 1H-Indole-3-acetic acid 2,4,5,6,7,alpha,alpha-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.