missing translation for 'onlineSavingsMsg'
Learn More

Indoine Blue, Thermo Scientific™

Product Code. 15402152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15402152 1 g 1g
10631273 5 g 5g
10181092 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15402152 Supplier Thermo Scientific Acros Supplier No. 365330010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4569-88-4
Molecular Formula C30H24ClN5O
Molecular Weight (g/mol) 506.01
MDL Number MFCD00011924
InChI Key SCMDRBZEIUMBBQ-UHFFFAOYSA-N
PubChem CID 9575841
SMILES [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Specifications

Infrared Spectrum Authentic
IUPAC Name 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Lambda Max 595 to 601nm (0.001% in methanol)
Formula Weight 506.01
Loss on Drying 10% max. (110°C, 1 h)
Quantity 1 g
Chemical Name or Material Indoine Blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.