missing translation for 'onlineSavingsMsg'
Learn More

Indocyanine Green, Analytical Reagent, MP Biomedicals

Product Code. 11478314 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg

Product Code. 11478314

Brand: MP Biomedicals 0521066750

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
MDL Number MFCD00013078
Synonym Cardiogreen, Foxgreen, IC Green
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
Molecular Weight (g/mol) 774.967
PubChem CID 132274068
ChEBI CHEBI:31696
Formula Weight 774.98
Color Green
Quantity 50 mg
Physical Form Powder
Chemical Name or Material Indocyanine Green
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.