missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Indocyanine green
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Product Type | Dye Reagents |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Synonym | Foxgreen ∣ IC Green |
| CAS | 3599-32-4 |
| Molecular Weight (g/mol) | 774.96 g/mol |
| Solubility | DMSO : 33.33 mg/mL (43.01 mM; Need ultrasonic) ∣ H2O : 5 mg/mL (6.45 mM; ultrasonic and warming and heat to 60°C) |
| Target | HBV |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction