missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Indocyanine green
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Indocyanine green is a near infrared (NIR) fluorescent dye operating in the 700-900 nm wavelength range. It also exhibits fluorescence emission in the shortwave infrared window (SWIR; 1,000-2,000 nm) which enables in vivo imaging, including intravital microscopy, non-invasive real-time imaging in blood and lymph vessels, and imaging of hepatobiliary clearance. Imaging of indocyanine green in SWIR shows increased sensitivity, contrast, and resolution of fine anatomical structures over NIR.
This product is supplied with a high degree of hydration and may contain residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Specifications
Specifications
| Chemical Name or Material | Sodium 4-(2-((1 E,3 E,5 E,7 Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2 H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1 H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate |
| CAS | 3599-32-4 |
| Quantity | 10 mg |
| Target | Fluorescent Dyes |
| Molecular Formula | C43H47N2O6S2Na |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction