missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water (I-Z)
Click to view available options
Display Text:
Industrial water
Low pressure water
Mineral water
Non-drinking water
Plant water
Polluted water
Primary circuit
Process water
Pure water
Purified water
Raw water
Return
River water
Secondary circuit
Soft water
Supply
Treated water
Waste water
Water
Well water
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Pack of 4
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
| Description | Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Primary circuit |
| Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
| Color | White on Green |
| Finish | Gloss |
| Format | Card |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Primary circuit |
| For Use With (Application) | Pipe and valve marking |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction