missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Water

Product Code. 15349415
Click to view available options
Display Text:
Boiler feed water
Brine Water
Canal Water
Chilled Water
Chlorated Water
City Water
Cold Water Return
Cold Water supply
Condensate
Condenser Water
Condenser Water Return
Condenser Water supply
Cooling Water
Decarbonatized Water
Deionized Water
Demineralised Water
Distilled Water
Drinking Water
Filtered Water
Heating
Heating Return
Heating Water
Heating supply
Hot Water
Hot water return
Industrial Water
Low pressure Water
Mineral Water
Non-drinking Water
Plant Water
Polluted Water
Process Water
Pure Water
Purified Water
Raw Water
Return
River Water
Secondary circuit
Soft Water
Supply
Treated Water
Waste Water
Water
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 15349415

Brand: Brady™ N009178

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Water - Cold water return
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color White on Green
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Cold Water Return
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 37 x 335 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Height (Metric) Character 16.5 mm
Alternate Part Number COLD WATER RETURN 355X37CARD-T1-P03
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.