missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads - Steam

Product Code. 15339485
Change view
Click to view available options
Display Text:
HP Steam
High Pressure Steam
Low Pressure Steam
Overheated Steam
Saturated Steam
Steam
Width (Metric):
250 mm
450 mm
Unit Size:
Pack of 2
Product Code. Display Text Width (Metric) unitSize
15339485 Steam 250 mm -
15359495 High Pressure Steam 250 mm -
15379495 High Pressure Steam 450 mm Pack of 2
15319505 HP Steam 250 mm -
15339505 HP Steam 450 mm Pack of 2
15319495 Low Pressure Steam 450 mm Pack of 2
15399485 Low Pressure Steam 250 mm -
15379505 Overheated Steam 250 mm -
15399505 Overheated Steam 450 mm -
15369505 Saturated Steam 450 mm Pack of 2
15349505 Saturated Steam 250 mm -
15359485 Steam 450 mm -
12 options
This item is not returnable. View return policy

Product Code. 15339485

Brand: Brady™ N009447

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Steam - Steam
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on Silver
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Steam
For Use With (Application) Pipe and valve marking
Labels per Sheet 4
Languages English
No. per Pack 4 Markers/Card
Quantity 4 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 26 x 250 mm
Length (English) 1.02 in.
Length (Metric) 26 mm
Width (English) 9.84 in.
Width (Metric) 250 mm
Height (Metric) Character 13.3 mm
Alternate Part Number STEAM 250X26CARD-T1-P14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.