missing translation for 'onlineSavingsMsg'
Learn More

Indium wire, 2.0mm (0.08in) dia, Puratronic™, 99.998+% (metals basis), Thermo Scientific Chemicals

Product Code. 11357107
Click to view available options
Quantity:
1 m
25 cm
5 m
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Quantity 1 m
Percent Purity ≥99.998%
Grade Puratronic™
Odor Odorless
Weight ≈22.95g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Indium wire, 2.0mm (0.08 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'