missing translation for 'onlineSavingsMsg'
Learn More

Indium wire, 0.25mm (0.01in) dia, Puratronic™, 99.998+% (metals basis)

Product Code. 11384018
Change view
Click to view available options
Quantity:
2.5 m
10 m
25 m
Unit Size:
10m
2.5m
25m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384018 2.5 m 2.5m
11304028 10 m 10m
15426325 25 m 25m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384018 Supplier Thermo Scientific Alfa Aesar Supplier No. 040421.G3
  • ONLINE EXCLUSIVE PRICE
    £117.00
    £105.65 / 2.5m
    Save £11.35
    9% Off
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

Form: Approx.0.36g/m | CAS: 7440-74-6 | In | 114.82 g/mol

As solders, thermal fuses, fusible alloys, and in semiconductor industry in InAs and InSb forms

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Appearance Silvery-white wire
Quantity 2.5 m
Odor Odorless
Assay Percent Range >99.998% (Metals basis)
Form Wire
Chemical Name or Material Indium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.