missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, 99.99% (metals basis)

Product Code. 11343438
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343438 100 g 100g
11353438 5 g 5g
11363438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343438 Supplier Thermo Scientific Alfa Aesar Supplier No. 036348.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50926-11-9 | InOSn | 249.527 g/mol

Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
Odor Odorless
Appearance White powder
Chemical Name or Material Indium tin oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.