missing translation for 'onlineSavingsMsg'
Learn More

Indium Tin eutectic ingot, 99.95% (metals basis)

Product Code. 11460490
Change view
Click to view available options
Quantity:
10 g
5x 10 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460490 5x 10 g 50g
11363417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460490 Supplier Thermo Scientific Alfa Aesar Supplier No. 018159.50
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indium Tin eutectic ingot
Melting Point 117°C
Avg. Mol. Wt. or Mol. Wt. Range In:Sn; 52:48 wt%
Quantity 5x 10 g
Form Ingot
Assay Percent Notes (metals basis)
MDL Number MFCD00144477
Percent Purity 99.99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.