missing translation for 'onlineSavingsMsg'
Learn More

Indium powder, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11371638
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
This item is not returnable. View return policy
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Melting Point 156.6°C
Boiling Point 2072°C
Density 7.31 g/mL
UN Number UN3089
Quantity 5 g
Formula Weight 114.82
Percent Purity 99.99%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Indium powder
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'