Learn More
Alfa Aesar™ Indium, plasma standard solution, Specpure™, In 10,000μg/mL
Brand: Alfa Aesar™ 014411.AE
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III
Quantity | 100mL |
---|
Chemical Identifiers
Indium plasma standard solution | |
Specpure™ | |
UN3264 |
In 10,000μg/mL | |
MFCD00134048 |
Specifications
Indium plasma standard solution | |
Specpure™ | |
In in 5% HNO3 | |
UN3264 |
In 10,000μg/mL | |
Liquid | |
MFCD00134048 | |
100mL |