missing translation for 'onlineSavingsMsg'
Learn More

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%

Product Code. 11339877
Change view
Click to view available options
Quantity:
2 g
10 g
25 g
Unit Size:
10g
25g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339877 25 g 25g
11349877 2 g 2g
11359877 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339877 Supplier Thermo Scientific Alfa Aesar Supplier No. 042229.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of transparent electrical conducting electrodes | CAS: 1312-43-2 | In2O3 | 277.63 g/mol

It is used in the preparation of transparent electrical conducting electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Physical Form Viscous Liquid
Quantity 25 g
UN Number UN1993
Flash Point 13°C (55°F)
Solubility Information Soluble in hexane and toluene; Insoluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈20 wt%
Chemical Name or Material Indium oxide, Tin doped, polymeric precursor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.