missing translation for 'onlineSavingsMsg'
Learn More

Indium ingot/button,≈31mm (1.2 in.) dia. x 5.3mm (0.21 in.) thick, 99.99% (metals basis), Thermo Scientific™

Product Code. 15434945
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
CAS 7440-74-6
Molecular Formula In
MDL Number MFCD00134048
Quantity 1 Pc.
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES [In]
IUPAC Name indium
Molecular Weight (g/mol) 114.82
PubChem CID 5359967
ChEBI CHEBI:30430
Percent Purity 99.99%
Odor Odorless
Weight ≈28g/piece
Form Pieces
Assay (metals basis)
Chemical Name or Material Indium ingot/button, ≈31mm (1.2 in.) dia. x 5.3mm (0.21 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'