missing translation for 'onlineSavingsMsg'
Learn More

Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11367028
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
This item is not returnable. View return policy
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 10 g
Percent Purity 99.999%
Odor Odorless
Form Ingot
Assay (metals basis)
Chemical Name or Material Indium ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'