missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) trifluoromethanesulfonate, 99% min

Product Code. p-7050971
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11319277

Brand: Thermo Scientific Alfa Aesar 040131.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128008-30-0 | C3F9InO9S3 | 562.008 g/mol

Indium(III) trifluoromethanesulfonate is used as a Lewis acid catalyst in organic synthesis and as a co-catalyst in organometallic catalysis. It is also used as a reactant in the preparation of stable indium bacteriochlorins and decahydroquinoline-type toxins through intramolecular hetero Diels-Alder reactions. Further, it is used in the synthesis of benzoxazoles through cyclocondensations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128008-30-0
Molecular Formula C3F9InO9S3
Molecular Weight (g/mol) 562.008
MDL Number MFCD00144478
InChI Key UCYRAEIHXSVXPV-UHFFFAOYSA-K
Synonym indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t
PubChem CID 2734680
IUPAC Name indium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN3261
Solubility Information Slightly soluble in hydrocarbons and diethyl ether. Insoluble in water
Formula Weight 562.02
Percent Purity ≤99%
Sensitivity Hygroscopic
Chemical Name or Material Indium(III) trifluoromethanesulfonate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.