missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfide, 99.98% (metals basis)

Product Code. 11366239
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366239 10 g 10g
11356239 2 g 2g
11376239 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366239 Supplier Thermo Scientific Alfa Aesar Supplier No. 044835.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12030-24-9 | In2S3 | 325.82 g/mol

Indium(III) sulfide acts as mid band gap semiconductor. It is an important material for optoelectronic and photovoltaic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-24-9
Molecular Formula In2S3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011061
InChI Key SIXIBASSFIFHDK-UHFFFAOYSA-N
IUPAC Name diindium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[In+3].[In+3]

Specifications

Melting Point 1050°C
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 325.83
Percent Purity 99.98%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium (III) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.