missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
46.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Indium(III) sulfate is used Chemical synthesis, hardening agent in gold electroplating baths. It is also used to prepare other indium containing substances such as copper indium selenide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 250°C (decomposition) |
Physical Form | Lump/Powder |
Quantity | 10 g |
Solubility Information | Soluble in water and ethanol. Insoluble in basic salts. |
Formula Weight | 517.83 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Assay | (metals basis) |
Chemical Name or Material | Indium(III) sulfate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'