missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11380979
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11380979

Brand: Thermo Scientific Alfa Aesar 010711.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13464-82-9 | In2O12S3 | 517.80 g/mol

Indium(III) sulfate is used Chemical synthesis, hardening agent in gold electroplating baths. It is also used to prepare other indium containing substances such as copper indium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13464-82-9
Molecular Formula In2O12S3
Molecular Weight (g/mol) 517.80
MDL Number MFCD00011059
InChI Key XGCKLPDYTQRDTR-UHFFFAOYSA-H
Synonym indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
PubChem CID 26044
IUPAC Name indium(3+);trisulfate
SMILES [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 250°C (decomposition)
Physical Form Lump/Powder
Quantity 50 g
Solubility Information Soluble in water and ethanol. Insoluble in basic salts.
Formula Weight 517.83 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium(III) sulfate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.