missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfate, anhydrous, 99.99% (metals basis)

Product Code. 11339237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339237 100 g 100g
11349237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339237 Supplier Thermo Scientific Alfa Aesar Supplier No. 040102.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13464-82-9 | In2O12S3 | 517.80 g/mol

Indium(III) sulfate, anhydrous is used in the preparation of indium or indium containing substances like copper indium selenide. It is employed to electroplate indium metal, as a hardening agent in gold electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13464-82-9
Molecular Formula In2O12S3
Molecular Weight (g/mol) 517.80
MDL Number MFCD00011059
InChI Key XGCKLPDYTQRDTR-UHFFFAOYSA-H
Synonym indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
PubChem CID 26044
SMILES [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Gray
Physical Form Solid
Quantity 100 g
Solubility Information Soluble in water.
IUPAC Name diindium(3+) trisulfate
Formula Weight 517.83
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.438 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) sulfate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.