missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) selenide, 99.99% (metals basis)

Product Code. 11349229
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349229 5 g 5g
11359229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349229 Supplier Thermo Scientific Alfa Aesar Supplier No. 088280.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12056-07-4 | In2Se3

Indium(III) selenide is used in photovoltaic devices. It is also used for optoelectronics and in solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12056-07-4
Molecular Formula In2Se3
MDL Number MFCD00016147

Specifications

Melting Point ∽890°C
Color Silver to Gray
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 466.52
Odor Odorless
Appearance Silver gray pieces
Density 5.67 g/cm3
Chemical Name or Material Indium(III) selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.