missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) perchlorate octahydrate, 99.9% (metals basis), Thermo Scientific™

£74.94

Chemical Identifiers

CAS 13465-15-1
MDL Number MFCD00150241
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371748
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011642.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13465-15-1
MFCD00150241

Specifications

800°C
Crystalline
UN1481
99.9%
Hygroscopic
Indium(III) perchlorate octahydrate
13465-15-1
MFCD00150241
557.30 (413.18 Anhydrous)
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

SDS
Documents

Documents

Product Certifications