missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) oxide, 99.999% (metals basis), Thermo Scientific Chemicals
62.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Finds application in semiconductors and can be used as a resistive element in integrated circuits. It is thermally stable and is suitable for glass, optic and ceramic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 850°C (volat.) |
Physical Form | Sintered Granules |
Quantity | 10 g |
Merck Index | 14,4952 |
Solubility Information | Insoluble in water. |
Formula Weight | 277.64 |
Percent Purity | 99.999% |
Odor | Odorless |
Density | 7.18 g/mL |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'