missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol

Product Code. 11445330
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445330 25 mL 25mL
11455330 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445330 Supplier Thermo Scientific Alfa Aesar Supplier No. 044562.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol

Precursor used in the preparation of indium oxide containing films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38218-24-5
Molecular Formula C9H21InO3
Molecular Weight (g/mol) 292.082
MDL Number MFCD00210318
InChI Key OVZUSPADPSOQQN-UHFFFAOYSA-N
Synonym triisopropoxyindium iii, indium iii isopropoxide, indium, m5-oxotetrakis m-2-propanolato tetrakis m3-2-propanolato pentakis 2-propanolato penta-9ci, 2-propanol, indium 3+ salt 9ci, acmc-20mnpq, indium tripropan-2-olate, indium 3+ ion tris propan-2-olate, indium 3+ tris propan-2-olate, indium iii isopropoxide, 5 w/v % solutionin 2-propanol, 99.9+%
PubChem CID 57346963
IUPAC Name indium(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]

Specifications

Color Colorless
Density 0.78 g/mL
Boiling Point 82°C
Flash Point 12°C (54°F)
Linear Formula In[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ketones and esters.
Formula Weight 292.08
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 5% w/v isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Indium (III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.