missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
10 g
Description
Indium(III) isopropoxide acts as a precursor for the preparation of indium oxide containing films. It finds application in nano-materials and thin films display. Furthermore, it is used in imaging catalysis and in preparation of sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | 145°C (decomposition) |
Color | White |
Linear Formula | In[OCH(CH3)2]3 |
Quantity | 0.5 g |
UN Number | UN3181 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in alcohols,ketones and esters. |
Formula Weight | 292.08 |
Percent Purity | 99.9% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'