Learn More
Indium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™
£852.00
Chemical Identifiers
| CAS | 13510-35-5 |
|---|---|
| Molecular Formula | I3In |
| Molecular Weight (g/mol) | 495.531 |
| MDL Number | MFCD00016151 |
| InChI Key | RMUKCGUDVKEQPL-UHFFFAOYSA-K |
| Synonym | indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis |
| PubChem CID | 83539 |
| IUPAC Name | triiodoindigane |
| SMILES | [In](I)(I)I |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemical Identifiers
| 13510-35-5 | |
| 495.531 | |
| RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
| 83539 | |
| [In](I)(I)I |
| I3In | |
| MFCD00016151 | |
| indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
| triiodoindigane |
Specifications
| 13510-35-5 | |
| MFCD00016151 | |
| indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
| RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
| triiodoindigane | |
| 83539 | |
| 99.999% | |
| Hygroscopic; Light sensitive | |
| (metals basis) |
| I3In | |
| UN3260 | |
| Soluble in water. | |
| [In](I)(I)I | |
| 495.531 | |
| 495.53 | |
| Ampule | |
| 4.69 g/mL | |
| Indium(III) iodide, Ultra dry |
Safety and Handling
GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335-H360
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 236-839-6
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.