missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, anhydrous, 99.999% (metals basis)

Product Code. 11381918
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11381918

Brand: Thermo Scientific Alfa Aesar 011854.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13510-35-5 | I3In | 495.531 g/mol

Indium(III) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-35-5
Molecular Formula I3In
Molecular Weight (g/mol) 495.531
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I

Specifications

Melting Point 210°C
Color Red to Yellow
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.994% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 495.53
Sensitivity Hygroscopic
Density 4.69
Chemical Name or Material Indium(III) iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.