missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, 99.998%, (trace metal basis)

Product Code. 10202992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202992 5 g Glass bottle 5g
10553421 1 g Glass bottle 1g
10711365 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10202992 Supplier Thermo Scientific Acros Supplier No. 316010050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13510-35-5 | InI3 | 495.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-35-5
Molecular Formula InI3
Molecular Weight (g/mol) 495.53
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I

Specifications

Melting Point 210.0°C
Color Brown-Yellow to Yellow
Physical Form Crystals or Powder
Quantity 5 g
Assay Percent Range 22.7 to 23.5% (In) (Complexometry)
Linear Formula InI3
Solubility Information Solubility in water: soluble
Formula Weight 495.53
Percent Purity 99.998%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) iodide
Show More Show Less
compliance-icons
Product Identifier
  • Indium(III) iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Indium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.