missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) hydroxide, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048885
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
20661-21-6
MFCD00011056
This item is not returnable. View return policy
20661-21-6
MFCD00011056

Indium(III) hydroxide, acts as a precursor to indium(III) oxide. It is used in batteries, glasses, ceramics, chemical reagents, semiconductor materials. It is also used used for manufacturing of panel displays and demist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150°C (in -H2O)
Physical Form Powder
Quantity 10 g
Solubility Information It is insoluble in water.
Formula Weight 165.84
Percent Purity 99.998%
Assay (metals basis)
Chemical Name or Material Indium(III) hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'