missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) fluoride, anhydrous, 96%

Product Code. 11379247
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379247 10 g 10g
11389247 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379247 Supplier Thermo Scientific Alfa Aesar Supplier No. 040118.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-52-0

Indium(III) fluoride is employed in the synthesis of non-oxide glasses. It promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-52-0
MDL Number MFCD00016149
Synonym indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride

Specifications

Boiling Point >1200°C
Melting Point ∼1170°C
Color White
Physical Form Powder
Quantity 10 g
Merck Index 14,4959
Solubility Information Slightly soluble in water (0;04g/100mL); Freely soluble in dilute acids
Formula Weight 171.82
Percent Purity 96%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.39 g/mL
Chemical Name or Material Indium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.