missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) Chloride, TRC

Product Code. 30403074
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403074 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403074 Supplier Toronto Research Chemicals Supplier No. TRCI5337251G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indium(III) Chloride
CAS 10025-82-8
Molecular Formula Cl3In
Quantity 1 g
Synonym Indium Chloride (InCl3), Indium Chloride, Indium Trichloride, Trichloroindium
InChI Formula InChI=1S/3ClH.In/h3*1H;/q;;;+3/p-3
SMILES Cl[In](Cl)Cl
IUPAC Name trichloroindigane
Molecular Weight (g/mol) 221.18
Formula Weight 219.8104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.