missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) Chloride Tetrahydrate, TRC

Product Code. 30394178
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394178 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394178 Supplier Toronto Research Chemicals Supplier No. TRCI5337552.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indium(III) Chloride Tetrahydrate
CAS 22519-64-8
Molecular Formula Cl3H8InO4
Quantity 2.5 g
InChI Formula InChI=1S/3ClH.In.4H2O/h3*1H;;4*1H2/q;;;+3;;;;/p-3
SMILES O.O.O.O.[Cl-].[Cl-].[Cl-].[In+3]
IUPAC Name indium(3+);trichloride;tetrahydrate
Molecular Weight (g/mol) 293.24
Formula Weight 291.8527
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.