missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride hydrate, 99.99% (metals basis)

Product Code. 11361928
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361928 50 g 50g
11381928 10 g 10g
11371928 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361928 Supplier Thermo Scientific Alfa Aesar Supplier No. 011859.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143983-91-9 | Cl3In | 221.17 g/mol

Indium(III) chloride hydrate is used for mild and selective cleavage of tert-butyldimethylsilyl ethers. It is a Lewis acid, and serves as a catalyst for many organic transformations. It also catalyzes the conversion of aldehydes to gem diacetates by chemoselective process. Lithium indium hydride, an in situ reducing agent, can be prepared from indium trichloride. It catalyzes reactions such as Friedel-Crafts acylations and Diels-Alder reactions. It also catalyzes Knoevenagel condensation between the barbituric acid and the aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143983-91-9
Molecular Formula Cl3In
Molecular Weight (g/mol) 221.17
MDL Number MFCD00011058
InChI Key PSCMQHVBLHHWTO-UHFFFAOYSA-K
IUPAC Name indium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[In+3]

Specifications

Color White
Physical Form Crystalline
Quantity 50 g
UN Number UN3260
Merck Index 14,4958
Solubility Information Soluble in water,diethyl ether,and THF.
Formula Weight 221.18 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Indium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.