missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, anhydrous, 99.999% (metals basis)

Product Code. p-7048886
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Unit Size:
10g
1g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331928 1 g 1g
11351928 10 g 10g
11341928 50 g 50g
11321928 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11331928 Supplier Thermo Scientific Alfa Aesar Supplier No. 011856.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-82-8

It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Merck Index 14,4958
Solubility Information It is very soluble in water and THF. It is sparingly soluble in alcohol and insoluble in ether.
Formula Weight 221.18
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Density 3.460 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.