missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) chloride, anhydrous, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.
Solubility
Soluble in water and tetrahydrofuran. Sparingly soluble in alcohol and insoluble in ether.
Notes
Hygroscopic. Incompatible with strong acids and strong oxidizing agents. The molten state of indium(III) chloride may conduct electricity.
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.
Solubility
Soluble in water and tetrahydrofuran. Sparingly soluble in alcohol and insoluble in ether.
Notes
Hygroscopic. Incompatible with strong acids and strong oxidizing agents. The molten state of indium(III) chloride may conduct electricity.
Specifications
Boiling Point | 800°C |
Melting Point | 586°C |
Quantity | 5 g |
UN Number | UN3260 |
Merck Index | 14,4958 |
Solubility Information | Soluble in water and tetrahydrofuran. Sparingly soluble in alcohol and insoluble in ether. |
Formula Weight | 221.18 |
Percent Purity | ≥98% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'