missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-4509689
50.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
10025-82-8
Cl3In
221.18
10025-82-8
Cl3In
221.18

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 586.0°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99.995%
Linear Formula InCl3
Merck Index 15,4998
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene
Formula Weight 221.18
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Indium(III) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'