Learn More
Indium(III) bromide, anhydrous, 99.99% (metals basis)
Description
Indium(III) bromide is used in organic synthesis as a water tolerant Lewis acid.It efficiently catalyzes the three-component coupling of β-keto esters, aldehydes and urea (or thiourea) to afford the corresponding dihydropyrimidinones. It is used as a catalyst to produce dithioacetals when unactivated alkynes react with thiols and fields such as optics and microelectronics that utilize semiconductor technology have wide uses for indium in high-performing solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼436°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder and/or Chunks |
| Quantity | 50 g |
| Linear Formula | InBr3 |
| Solubility Information | Soluble in water. |
| Formula Weight | 354.55 |
| Percent Purity | 99.99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
- Indium(III) bromide, anhydrous
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Indium bromide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.