missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) chloride, anhydrous, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 11389217
Click to view available options
Quantity:
0.5 g
5 g
25 g
This item is not returnable. View return policy
13465-10-6
Cl3In
221.17
MFCD00148875
PSCMQHVBLHHWTO-UHFFFAOYSA-K
indium(3+) trichloride
[Cl-].[Cl-].[Cl-].[In+3]
This item is not returnable. View return policy
13465-10-6
Cl3In
221.17
MFCD00148875
PSCMQHVBLHHWTO-UHFFFAOYSA-K
indium(3+) trichloride
[Cl-].[Cl-].[Cl-].[In+3]

Indium chloride is a lewis acid catalyst in organic reactions such as Friedel-Crafts acylations and Diels-Alder reactions and it is used in high-performing solar cells such as optics and microelectronics. It applications finds in semiconductor technology, producing phosphor,low pressure sodium lamp and produce high purity chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Boiling Point 608°C
Melting Point ∼225°C
Physical Form Crystalline
Quantity 0.5 g
UN Number UN3260
Beilstein EM>13,
Solubility Information Soluble in water and THF.
Formula Weight 150.27
Percent Purity 99.995%
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 4.19 g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'