missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) chloride, anhydrous, 99.995% (metals basis)

Product Code. 11399217
Change view
Click to view available options
Quantity:
0.5 g
5 g
25 g
Unit Size:
0.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399217 5 g 5g
11389217 0.5 g 0.5g
11309227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399217 Supplier Thermo Scientific Alfa Aesar Supplier No. 040033.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-10-6 | ClIn | 150.27 g/mol

Indium chloride is a lewis acid catalyst in organic reactions such as Friedel-Crafts acylations and Diels-Alder reactions and it is used in high-performing solar cells such as optics and microelectronics. It applications finds in semiconductor technology, producing phosphor,low pressure sodium lamp and produce high purity chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-10-6
Molecular Formula ClIn
Molecular Weight (g/mol) 150.27
MDL Number MFCD00148875
InChI Key APHGZSBLRQFRCA-UHFFFAOYSA-M
IUPAC Name λ¹-indiganyliumylidene chloride
SMILES [Cl-].[In+]

Specifications

Boiling Point 608°C
Melting Point ∼225°C
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Beilstein EM>13,
Solubility Information Soluble in water and THF.
Formula Weight 150.27
Percent Purity 99.995%
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 4.19 g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) chloride, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Indium chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Indium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.