missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11378168
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378168 5 g 5g
11388168 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378168 Supplier Thermo Scientific Alfa Aesar Supplier No. 039388.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14280-53-6
MDL Number MFCD00064630
Synonym indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci

Specifications

Boiling Point 662°C
Melting Point 220°C
Physical Form Beads
Quantity 5 g
UN Number UN3260
Formula Weight 194.73
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Moisture Sensitive
Density 4.96g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.