missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11340108
Click to view available options
Quantity:
5 g
25 g
14280-53-6
InBr
MFCD00064630
indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci
14280-53-6
InBr
MFCD00064630
indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 662°C
Melting Point 220°C
Physical Form Polycrystalline Lump
Quantity 5 g
UN Number UN3260
Formula Weight 194.73
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Sensitivity Moisture sensitive
Density 4.96 g/mL
Assay (metals basis)
Chemical Name or Material Indium (I) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'