missing translation for 'onlineSavingsMsg'
Learn More

Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386608
Click to view available options
Quantity:
50 x 50 mm
50 x 100 mm
100 x 150 mm
150 x 300 mm
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]
7440-74-6
In
114.82
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
5359967
CHEBI:30430
indium
[In]

Form: approx. 4.6g/50x50mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver-White
Appearance Silvery-white foil
Assay Percent Range 99.99% (Metals basis)
Quantity 150 x 300 mm
Odor Odorless
Form Foil
Chemical Name or Material Indium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'