missing translation for 'onlineSavingsMsg'
Learn More

Indium arsenide, 99% (metals basis)

Product Code. 11358077
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358077 2 g 2g
11368077 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11358077 Supplier Thermo Scientific Alfa Aesar Supplier No. 022651.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

In semiconductor electronics | CAS: 1303-11-3 | AsIn | 189.74 g/mol

Indium arsenide is used in semiconductor electronics, construction of infrared detectors, terahertz radiation source, as it is a strong Photo-dember emitter, superior electron mobility and velocity, in high-power applications detector, diode lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-11-3
Molecular Formula AsIn
Molecular Weight (g/mol) 189.74
MDL Number MFCD00016144
InChI Key RPQDHPTXJYYUPQ-UHFFFAOYSA-N
Synonym indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
PubChem CID 91500
IUPAC Name indiganylidynearsane
SMILES [As]#[In]

Specifications

Melting Point 936°C
Physical Form Pieces
Quantity 2 g
Linear Formula InAs
UN Number UN1557
Merck Index 14,4949
Solubility Information Insoluble in water.
Formula Weight 189.74
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Indium arsenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.