missing translation for 'onlineSavingsMsg'
Learn More

Indium antimonide, Electronic Grade, 99.99% (metals basis)

Product Code. 11399269
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399269 50 g 50g
11379269 2 g 2g
11389269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399269 Supplier Thermo Scientific Alfa Aesar Supplier No. 088460.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In semiconductor electronics, as an infrared detector and filter | CAS: 1312-41-0 | InSb | 236.578 g/mol

Indium antimonide is used in semiconductor electronics, as an infrared detector and filter. Used in thermal imaging cameras, FLIR systems, infrared homing missile guidance systems, photo-Dember emitter and in infrared astronomy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-41-0
Molecular Formula InSb
Molecular Weight (g/mol) 236.578
MDL Number MFCD00016146
InChI Key WPYVAWXEWQSOGY-UHFFFAOYSA-N
PubChem CID 3468413
IUPAC Name indiganylidynestibane
SMILES [In]#[Sb]

Specifications

Melting Point 535°C
Physical Form Pieces
Quantity 50 g
UN Number UN1549
Merck Index 14,4948
Solubility Information Insoluble in water.
Formula Weight 236.57
Percent Purity 99.99%
Grade Electronic
Odor Odorless
Density 5.75 g/mL at (m.p.)
Assay (metals basis)
Chemical Name or Material Indium antimonide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.