missing translation for 'onlineSavingsMsg'
Learn More

Indisetron, TRC

Product Code. 30390779
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390779 1 mg -
30404522 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390779 Supplier Toronto Research Chemicals Supplier No. TRCI5325001MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141549-75-9
Molecular Formula C17H23N5O
Molecular Weight (g/mol) 313.4
Synonym N-[(7-endo)-3,9-Dimethyl-3,9-diazabicyclo[3.3.1]non-7-yl]-1H-indazole-3-carboxamide, endo-N-(3,9-Dimethyl-3,9-diazabicyclo[3.3.1]non-7-yl)-1H-indazole-3-carboxamide
IUPAC Name N-[(1S,5R)-3,9-dimethyl-3,9-diazabicyclo[3.3.1]nonan-7-yl]-1H-indazole-3-carboxamide
SMILES CN1C[C@H]2C[C@H](C[C@@H](C1)N2C)NC(=O)c3n[nH]c4ccccc34

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H23N5O/c1-21-9-12-7-11(8-13(10-21)22(12)2)18-17(23)16-14-5-3-4-6-15(14)19-20-16/h3-6,11-13H,7-10H2,1-2H3,(H,18,23)(H,19,20)/t11-,12-,13+
Formula Weight 313.1903
Chemical Name or Material Indisetron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.